missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118863
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SOS2.
SOS2 promotes the exchange of Ras-bound GDP by GTP.
Specifications
| SOS2 | |
| Polyclonal | |
| Unconjugated | |
| SOS2 | |
| mSOS-2; NS9; son of sevenless 1; son of sevenless homolog 2; son of sevenless homolog 2 (Drosophila); SOS Ras/Rho guanine nucleotide exchange factor 2; Sos1; Sos2; SOS-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6655 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07890 | |
| SOS2 | |
| A synthesized peptide derived from human SOS2(Accession Q07890), corresponding to amino acid residues R630-V680. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction