missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SOS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15984104
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15984104 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15984104 Supplier Invitrogen™ Supplier No. 720413

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Pig, Cat, Mouse.

SOS2 promotes the exchange of Ras-bound GDP by GTP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOS2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene SOS2
Gene Accession No. Q07890
Gene Alias mSOS-2; NS9; son of sevenless 1; son of sevenless homolog 2; son of sevenless homolog 2 (Drosophila); SOS Ras/Rho guanine nucleotide exchange factor 2; Sos1; Sos2; SOS-2
Gene Symbols SOS2
Host Species Rabbit
Immunogen Peptides corresponding to human SOS2 [1) aa1152-aa1170 2) aa1184-aa1203].
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6655
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.