missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOST/Sclerostin Antibody (2), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89575B
This item is not returnable.
View return policy
Description
SOST/Sclerostin Monoclonal antibody specifically detects SOST/Sclerostin in Human samples. It is validated for ELISA
Specifications
| SOST/Sclerostin | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 2 | |
| ELISA | |
| sclerostin, VBCHsclerosteosis | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human SOST/Sclerostin (NP_079513.1; Gln 24-Tyr 213). | |
| 0.1 mL | |
| Signal Transduction | |
| 50964 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction