missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24486UV
This item is not returnable.
View return policy
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Equine, Opossum, Primate, Xenopus samples. It is validated for Western Blot.
Specifications
| SOX1 | |
| Polyclonal | |
| Western Blot | |
| SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SOX1 | |
| A portion of amino acids 100-150 of human SOX1 was used as the immunogen. | |
| 0.1 mL | |
| Stem Cells | |
| Human, Mouse, Equine, Opossum, Primate, Xenopus species | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction