missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Antibody (2447A), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB28642G
This item is not returnable.
View return policy
Description
SOX10 Monoclonal antibody specifically detects SOX10 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| SOX10 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6663 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry, CyTOF | |
| 2447A | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| E. coli-derived recombinant human SOX10, Met1-Ala118, Accession # P56693 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction