Learn More
SOX10 Antibody (2447A), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB28642CL1
This item is not returnable.
View return policy
Description
SOX10 Monoclonal antibody specifically detects SOX10 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| SOX10 | |
| Monoclonal | |
| CoraFluor 1 | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| E. coli-derived recombinant human SOX10, Met1-Ala118, Accession # P56693 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 2447A | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 6663 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |