missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Antibody (2447A), DyLight 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB28642K
This item is not returnable.
View return policy
Description
SOX10 Monoclonal antibody specifically detects SOX10 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| SOX10 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6663 | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 2447A | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| E. coli-derived recombinant human SOX10, Met1-Ala118, Accession # P56693 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction