missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18510B
This item is not returnable.
View return policy
Description
SOX10 Polyclonal antibody specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SOX10 | |
| Polyclonal | |
| Western Blot | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| Recognizes region between residues Between between 416 and 466.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6663 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion