missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX10 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18510G
This item is not returnable.
View return policy
Description
SOX10 Polyclonal antibody specifically detects SOX10 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SOX10 | |
| Polyclonal | |
| Western Blot | |
| MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of | |
| Recognizes region between residues Between between 416 and 466.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6663 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction