missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX10 Monoclonal Antibody (1-11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544626
This item is not returnable.
View return policy
Description
SOX10 Monoclonal Antibody for Western Blot, IHC (P)
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. This protein may act as a transcriptional activator after forming a protein complex with other proteins. It acts as a nucleocytoplasmic shuttle protein and is important for neural crest and peripheral nervous system development. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Specifications
| SOX10 | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 0.2% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q04888 | |
| SOX10 | |
| Recombinant protein within human SOX10 aa 80-310. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1-11 | |
| Unconjugated | |
| SOX10 | |
| cls; cls/sox10; colorless; colourless; colourless/sox10; DOM; dominant megacolon; dominant megacolon, mouse, human homolog of; golas; gos; HMG-box transcription factor Sox10; MGC15649; PCWH; Protein SOX-21; SOX10; Sox-10; sox10 protein; sox10b; Sox21; SOX9; SRY; SRY (sex determining region Y)-box 10; SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal); SRY box 10; SRY-box 10; SRY-box containing gene 10; SRY-box containing protein 10; SRY-box transcription factor 10; SRY-related HMG-box gene 10; transcription factor SOX-10; Transcription factor SOX-M; WS2E; WS4; WS4C; zgc:100757 | |
| Mouse | |
| Protein A | |
| RUO | |
| 20665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction