Learn More
SOX10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10422-1-AP-150UL
 View more versions of this product
Product Code. 16816803
  • € 410.00 / 150µL
Description
SOX10 is a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. SOX10 plays a central role in developing and maturing of glia. Specifically, SOX10 activates expression of myelin genes (such as DUSPP15 and MYRF) during oligodendrocyte (OL) maturation; thus, playing a central role in oligodendrocyte maturation and CNS myelination. Once induced MYRF cooperates with SOX10 to implement the myelination program. SOX10 is also a transcriptional activator of MITF acting synergistically with PAX3. Mutations in this gene are associated with Waardenburg-Shah and Waardenburg-Hirschsprung disease.
Specifications
Specifications
SOX10
Polyclonal
Unconjugated
SOX10
DOM, SOX10, Transcription factor SOX 10, WS2E, WS4
Rabbit
Antigen Affinity Chromatography
RUO
20665, 29361, 6663
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O55170, P56693, Q04888
SOX10
SOX10 Fusion Protein Ag0673
150 μL
Primary
Human, Mouse, Rat