missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX11 Antibody (SOX11/3235R), Alexa Fluor™ 750, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08594AF750
This item is not returnable.
View return policy
Description
SOX11 Monoclonal antibody specifically detects SOX11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SOX11 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| SRY (sex determining region Y)-box 11, SRY (sex-determining region Y)-box 11, SRY-related HMG-box gene 11, transcription factor SOX-11 | |
| Recombinant fragment (around aa300-400) of human SOX11 protein (Uniprot: P35716 ) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| SOX11/3235R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 6664 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction