missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX11 Recombinant Rabbit Monoclonal Antibody (5K1J5)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551319
This item is not returnable.
View return policy
Description
Immunogen sequence: MVQQAESLEA ESNLPREALD TEEGEFMACS PVALDESDPD WCKTASGHIK RPMNAFMVWS KIERRKIMEQ SPDMHNAEIS KRLGKRWKML KDSEKIPFIR.
This intronless gene encodes a member of the SOX family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may function in the developing nervous system and play a role in tumorigenesis.
Specifications
| SOX11 | |
| Recombinant Monoclonal | |
| 0.51 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P0C1G9, P35716, Q7M6Y2 | |
| SOX11 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 5K1J5 | |
| Unconjugated | |
| SOX11 | |
| 1110038H03Rik; 6230403H02Rik; AI836553; end1; MRD27; Sox11; Sox-11; SRY; SRY (sex determining region Y)-box 11; SRY (sex-determining region Y)-box 11; SRY-box 11; SRY-box containing gene 11; SRY-box transcription factor 11; SRY-related HMG-box gene 11; transcription factor SOX-11 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20666, 6664, 84046 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction