missing translation for 'onlineSavingsMsg'
Learn More

SOX13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. p-7208385 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16838744 20 μL 20µL
16828744 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16838744 Supplier Proteintech Supplier No. 189021AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. It has also been determined to be a type-1 diabetes autoantigen, also known as islet cell antibody 12.

Specifications

Antigen SOX13
Applications Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SOX13
Gene Accession No. Q04891, Q9UN79
Gene Alias ICA12, Islet cell antigen 12, Sox 13, SOX13, Transcription factor SOX 13
Gene Symbols Sox13
Host Species Rabbit
Immunogen SOX13 Fusion Protein Ag13537
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20668, 289026, 9580
Target Species Mouse, Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.