missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX17 Antibody (OTI2B6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74299F
This item is not returnable.
View return policy
Description
SOX17 Monoclonal specifically detects SOX17 in Human, Mouse, Rat, Canine samples. It is validated for Immunohistochemistry.
Specifications
| SOX17 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SOX17 | |
| Recombinant protein expressed in E.coli corresponding to amino acids 177-414 of human SOX17. | |
| 0.1 mL | |
| 64321.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI2B6 | |
| Immunohistochemistry | |
| FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction