missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA572815
This item is not returnable.
View return policy
Description
SOX17 Polyclonal Antibody for ELISA, IHC, Western Blot
SOX17 (transcription factor SOX-17) belongs to the SRY-related HMB box family of transcription factors. It acts as a transcription regulator that binds target promoter DNA and bends the DNA. It binds to the sequences 5'-AACAAT-'3 or 5'-AACAAAG-3'. SOX17 modulates transcriptional regulation via WNT3A and inhibits Wnt signaling. It promotes degradation of activated CTNNB1. SOX17 plays an essential role in the regulation of embryonic development and is required for normal looping of the embryonic hear tube and for normal development of the definitive gut endoderm. Mutations in the gene can result in vesicoureteral reflux.
Specifications
| SOX17 | |
| Polyclonal | |
| Unconjugated | |
| SOX17 | |
| HMG transcription factor SOX17; HMG-box transcription factor Sox17; Sox17; Sox-17; SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-box containing gene 17; SRY-box transcription factor 17; SRY-related HMG-box transcription factor SOX17; transcription factor SOX-17; transcription factor SOX-17; LOW QUALITY PROTEIN: transcription factor SOX-17; VUR3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20671, 312936, 64321 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q61473, Q9H6I2 | |
| SOX17 | |
| SOX17 antibody for a 16 amino acid peptide near the center of human SOX17. The immunogen is within amino acids 240 - 290 of SOX17. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction