missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX18 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41289JF646
This item is not returnable.
View return policy
Description
SOX18 Polyclonal antibody specifically detects SOX18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| SOX18 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 54345 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| Janelia Fluor 646 | |
| HLTS, SRY (sex determining region Y)-box 18, SRY-box 18, transcription factor SOX-18 | |
| Antibody was raised against a 17 amino acid peptide near the amino terminus of human SOX18. The immunogen is located within amino acids 40 - 90 of SOX18. Amino Acid Squence: ASPPSPQRSPPRSPEPG | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction