missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Antibody (245610R), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2018RD
This item is not returnable.
View return policy
Description
SOX2 Monoclonal specifically detects SOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.
Specifications
| SOX2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry | |
| 245610R | |
| Western Blot, Intracellular Staining by Flow Cytometry | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| E.coli-derived recombinant human SOX2, Gly135-Met317, Accession # P48431 | |
| 0.1 mL | |
| Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 6657 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction