missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX2 Rabbit anti-Human, Mouse, Clone: SOX2/3811R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07893-100ug
This item is not returnable.
View return policy
Description
SOX2 Monoclonal specifically detects SOX2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.
Specifications
| SOX2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| Recombinant fragment (within aa176-305) of human SOX2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cellular Markers, Core ESC Like Genes, Neuroscience, Sensory Systems, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators, Vision | |
| 6657 | |
| Store at 4°C. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| SOX2/3811R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction