Learn More
SOX30 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-35189-100ul
 Afficher plus de versions de ce produit
Code produit. 30230412
  • € 550.00 / 100µL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
SOX30 Polyclonal antibody specifically detects SOX30 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Spécification
Spécification
SOX30
Polyclonal
Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200
Sox30 protein type II, SRY (sex determining region Y)-box 30, transcription factor SOX-30
Recombinant fusion protein containing a sequence corresponding to amino acids 604-753 of human SOX30 (NP_848511.1).,, Sequence:, FGTPPRFSFHHPYFLPGPHYFPSSTCPYSRPPFGYGNFPSSMPECLSYYEDRYPKHEGIFSTLNRDYSFRDYSSECTHSENSRSCENMNGTSYYNSHSHSGEENLNPVPQLDIGTLENVFTAPTSTPSSIQQVNVTDSDEEEEEKVLRDL
100 μL
Primary
Human, Mouse, Rat
Purified
ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
11063
Store at -20°C. Avoid freeze-thaw cycles.