missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 Antibody (PCRP-SOX4-1D6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14180CL1
This item is not returnable.
View return policy
Description
SOX4 Monoclonal antibody specifically detects SOX4 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Protein Array,Immunocytochemistry/Immunofluorescence
Specifications
| SOX4 | |
| Monoclonal | |
| CoraFluor 1 | |
| ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
| Recombinant fragment (around aa45-130) of human SOX4 protein (Uniprot: Q06945) | |
| 0.1 mL | |
| Stem Cell Markers, Stem Cells | |
| 6659 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| PCRP-SOX4-1D6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction