missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX4 Mouse anti-Human, Clone: PCRP-SOX4-1D6, Novus Biologicals™
Mouse Monoclonal Antibody
€ 276.00 - € 547.00
Specifications
| Antigen | SOX4 |
|---|---|
| Clone | PCRP-SOX4-1D6 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388874
|
Novus Biologicals
NBP3-13777-20UG |
20 μg |
€ 276.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18366856
|
Novus Biologicals
NBP3-13777-100UG |
100 μg |
€ 547.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX4 Monoclonal antibody specifically detects SOX4 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| SOX4 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Stem Cell Markers, Stem Cells | |
| 10 mM PBS with 0.05% BSA | |
| ecotropic viral integration site 16, EVI16, SRY (sex determining region Y)-box 4, SRY-related HMG-box gene 4, transcription factor SOX-4 | |
| Recombinant fragment (around aa45-130) of human SOX4 protein (Uniprot: Q06945) | |
| Primary | |
| Store at 4°C. |
| PCRP-SOX4-1D6 | |
| Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q06945 | |
| 6659 | |
| IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title