missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX5 Antibody (OTI2A8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74301H
This item is not returnable.
View return policy
Description
SOX5 Monoclonal specifically detects SOX5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SOX5 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| SOX5 | |
| Full length recombinant protein of human SOX5 (NP_008871) produced in HEK293T cells. | |
| 0.1 mL | |
| 6660.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2A8 | |
| Western Blot | |
| L-SOX5, MGC35153, SRY (sex determining region Y)-box 5, transcription factor SOX-5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction