Learn More
SOX6 Antibody (CL5690) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61423AF750
Product Code. 30301244
  • € 737.00 / 0.10mL
Description
SOX6 Monoclonal antibody specifically detects SOX6 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOX6
Monoclonal
Alexa Fluor 750
HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6
This antibody was developed using a synthetic peptide derived from P35712, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL5690
50mM Sodium Borate
Mouse
Protein A purified
RUO
55553
Store at 4⪚C in the dark.
IgG2b