missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX6 Antibody (CL5690), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61423-25ul
This item is not returnable.
View return policy
Description
SOX6 Monoclonal specifically detects SOX6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SOX6 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
| SOX6 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: NLDTFEHGGGHSYNHKQIE | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL5690 | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| HSSOX6, SOXD, SRY (sex determining region Y)-box 6, SRY-box containing gene 6, transcription factor SOX-6 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 55553 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction