missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX6 Monoclonal Antibody (OTI5E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808936
This item is not returnable.
View return policy
Description
Transcriptional activator. Binds specifically to the DNA sequence 5′-AACAAT-3′. Plays a key role in several developmental processes, including neurogenesis and skeleton formation.Specifications
| SOX6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P35712 | |
| SOX6 | |
| Human recombit protein fragment corresponding to amino acids 1-244 of human SOX6 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5E4 | |
| Unconjugated | |
| SOX6 | |
| HSSOX6, SOXD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55553 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction