Learn More
SOX6 Monoclonal Antibody (OTI5E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808936S
Encompass_Preferred
 View more versions of this product
Product Code. 17540724
  • € 242.00 / 30µL
Description
SOX6 Monoclonal antibody specifically detects SOX6 in Human samples. It is validated for Western Blot
Specifications
Specifications
SOX6
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q12792
Mouse
Affinity chromatography
RUO
5756
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5E4
Unconjugated
TWF1
AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6
Human recombinant protein fragment corresponding to amino acids 140-384 of human TWF1 produced in E.coli.