Learn More
Invitrogen™ SOX6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA110882
Description
PA1-10882 detects SOX6 in human and mouse samples. This antibody is predicted to recognize rat and chimapanzee SOX6 based on 100% sequence homology. PA1-10882 has been successfully used in Western blot procedures. PA1-10882 immunogen is a synthetic peptide corresponding to residues 780-796 of human and mouse SOX6.
The SOX6 gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors, which are known to play vital roles in the regulation of developmental processes and cell fate determination. SOX6, along with its close relatives SOX5 and SOX13, constitutes the SOXD subfamily, which is involved in the regulation of the development of the central nervous system, skeletal system, and cartilage. It functions by binding to specific DNA sequences and modulating the expression of target genes necessary for these developmental processes. SOX6 has been implicated in various functions, such as promoting cartilage formation by acting in conjunction with SOX9 to drive chondrogenesis. In adult neural stem cells, SOX6 is essential for the activation of radial glial-like cells and the generation of new neurons, emphasizing its role in both embryonic development and adult tissue maintenance. The dysregulation of SOX6 has been associated with developmental disorders and its potential involvement in pathologies like cancer, where it influences cell differentiation and proliferation pathways.
Specifications
| SOX6 | |
| Polyclonal | |
| PBS with no preservative; pH 7.4 | |
| P35712, P40645 | |
| SOX6 | |
| Synthetic peptide corresponding to residues 780-796 of human and mouse SOX6. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| SOX6 | |
| AI987981; HSSOX6; SOX6; Sox-6; SOXD; SOX-LZ; SRY (sex determining region Y)-box 6; SRY box 6; SRY-box 6; SRY-box containing gene 6; transcription factor SOX-6 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 20679, 55553 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.