missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX7 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-26090R
This item is not returnable.
View return policy
Description
SOX7 Polyclonal antibody specifically detects SOX7 in Mouse samples. It is validated for Western Blot
Specifications
| SOX7 | |
| Polyclonal | |
| Western Blot | |
| MGC10895, SOX7 transcription factor, SRY (sex determining region Y)-box 7, transcription factor SOX-7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse SOX7 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 83595 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction