missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX7 Monoclonal Antibody (288610)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561202
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may play a role in tumorigenesis. A similar protein in mice is involved in the regulation of the wingless-type MMTV integration site family (Wnt) pathway.
Specifications
| SOX7 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9BT81 | |
| Sox7 | |
| E. coli-derived recombinant human SOX7, Ser249-Ala345. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 288610 | |
| Unconjugated | |
| Sox7 | |
| MGC10895; mSOX7; SOX7; Sox-7; SRY (sex determining region Y)-box 7; SRY box 7; SRY-box 7; SRY-box containing gene 7; SRY-box transcription factor 7; transcription factor SOX-7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 83595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction