missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOX7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582375
This item is not returnable.
View return policy
Description
Immunogen sequence: SPLHCSHPLG SLALGQSPGV SMMSPVPGCP PSPAYYSPAT YHPLHSNLQA HLGQLSPPPE HPGFDALDQL SQVELLGDMD RNEFDQYLNT PGHPDSATGA MALSGHVPVS QVTPTGPTET SLISVLADAT ATYYNSYS Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.
This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. The protein may play a role in tumorigenesis. A similar protein in mice is involved in the regulation of the wingless-type MMTV integration site family (Wnt) pathway.
Specifications
| SOX7 | |
| Polyclonal | |
| Unconjugated | |
| Sox7 | |
| MGC10895; mSOX7; SOX7; Sox-7; SRY (sex determining region Y)-box 7; SRY box 7; SRY-box 7; SRY-box containing gene 7; SRY-box transcription factor 7; transcription factor SOX-7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83595 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BT81 | |
| Sox7 | |
| Recombinant protein corresponding to Human SOX7. Recombinant protein control fragment (Product #RP-89402). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction