missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX9 Antibody (CL0639), Novus Biologicals™
Mouse Monoclonal Antibody
€ 289.00 - € 573.00
Specifications
| Antigen | SOX9 |
|---|---|
| Clone | CL0639 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18629786
|
Novus Biologicals
NBP2-52943-25ul |
25 μL |
€ 289.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18058435
|
Novus Biologicals
NBP2-52943 |
0.1 mL |
€ 573.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SOX9 Monoclonal antibody specifically detects SOX9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDownSpecifications
| SOX9 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SQRTHIKTEQLSPSHYSEQQQHSPQQIAYSPFNLPHYSPSYPPITRSQYDYTDHQNSSSYYSHAAGQGTGLYSTFTYMNPAQRPMYTPIADTSGVPSIPQTHSPQHWEQPVYTQLTR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL0639 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| Mouse | |
| Cancer, Stem Cell Markers | |
| PBS (pH 7.2), 40% Glycerol | |
| 6662 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title