missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOX9 Antibody (SOX9/2287R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08592AF405
This item is not returnable.
View return policy
Description
SOX9 Monoclonal antibody specifically detects SOX9 in Human samples. It is validated for ELISA
Specifications
| SOX9 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| campomelic dysplasia, autosomal sex-reversal, CMD 1, CMD1, CMPD1, SRA1SRY (sex-determining region Y)-box 9 protein, SRY (sex determining region Y)-box 9, SRY-related HMG-box, gene 9, transcription factor SOX-9 | |
| Recombinant human full-length SOX9 protein (Uniprot: P48436) | |
| 100 μg | |
| Cancer, Stem Cell Markers | |
| 6662 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| SOX9/2287R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction