Learn More
Bel-Art™ SP Scienceware™ Full-View Test Tube Supports
Versatile polypropylene racks feature pins for support or draining of test tubes in a compact space
€ 27.93
Product Code | Brand | Positions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Positions | Price | Quantity & Availability | |||||
10505074
|
Bel-Art™
H18911-0000 |
50 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Tubes can be stored upright, full or empty, or inverted over pins for draining
- Steam autoclavable at 121°C (250°F)
Specifications
Autoclavable | |
Polypropylene |