Learn More
Bel Art™ Heavy-Duty Test-Tube Racks
Product Code. 15203109 Shop All SP Scienceware Products
Click to view available options
Positions:
15
18
32
50
72
Holds:
Tubes (10 to 13 mm)
Tubes (13 to 16 mm)
Tubes (16 to 20 mm)
Tubes (20 to 25 mm)
Tubes (25 to 30 mm)
Array:
3 x 5
3 x 6
4 x 8
5 x 10
6 x 12
Unit Size:
Each
Product Code. 15203109
Brand: Bel Art™ F188570000
Only null left
Add to basket
Edge
Add to basket
Heavy-duty 3-tier test tube racks have hand grips in the end plates for easy transport.
  • Stackable, compact and steam autoclavable at 121°C (250°F) for durability in the lab
  • End plates have hand grips for easy transport
  • Base tier has rounded wells with drain holes, and the top tier has lettered and numbered rows
  • Available in different sizes
  • 8.3 x 4.3 in. (21.3 x 11 cm)
Specifications
Product Type Test Tube Rack
Description Test Tube Rack
Array 4 x 8
Autoclavable Autoclavable
Color Translucent
Holds Tubes (16 to 20 mm)
Material Polypropylene
Positions 32
Shape Rectangular
Shelves 3 Tiers
Stackable No
Sterility Non-sterile
For Use With (Equipment) 16-20mm Test Tubes
Quantity 1 Each
Dimensions (L x W x H) 21.3 x 11 x 12.7 cm
Length (Metric) 21.3 cm
Width (Metric) 11 cm
Height (Metric) 12.7 cm
Diameter (Metric) Opening 20 mm