missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Wide Mouth Color-Code Safety Labeled Wash Bottles
Click to view available options
Capacity:
16 oz., 500 mL
For Use With (Application):
Ethyl Acetate
Methyl Ethyl Ketone
Description
Includes
Labeling with Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information
Easy-to-read labeling printed bottle promoting safe working environment
- Wide mouth 53mm (2 in.) bottle for easy refilling
- Made from solvent-resistant low-density polyethylene
- Polypropylene closure with leakproof design
- Pictogram instructions promote universal safety regardless of setting
Use LYOB only with solvents compatible with LDPE.
Compliance
Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Specifications
Capacity | 16 oz., 500 mL |
Closure Material | Polypropylene |
Closure Size | 53 mm |
Closure Type | Polypropylene Closure |
Through the Closure Dispensing | Yes |
Product Type | Wash Bottle |
Capacity (Metric) | 500 mL |
Capacity (English) | 16 oz. |
Color | Clear |
For Use With (Application) | Methyl Ethyl Ketone |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'