missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP100 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97407G
This item is not returnable.
View return policy
Description
SP100 Polyclonal specifically detects SP100 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SP100 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 6672 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| DKFZp686E07254, FLJ00340, FLJ34579, lysp100b, nuclear antigen Sp100, nuclear autoantigen Sp-100, Nuclear dot-associated Sp100 protein, SP100 nuclear antigen, SP100-HMG nuclear autoantigen, Speckled 100 kDa | |
| Produced in rabbits immunized with E. coli-derived Human SP100 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction