missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP100 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546706
This item is not returnable.
View return policy
Description
Peptide sequence: IIVISSEDSE GSTDVDEPLE VFISAPRSEP VINNDNPLES NDEKEGQEAT.
This gene encodes a subnuclear organelle and major component of the PML (promyelocytic leukemia)-SP100 nuclear bodies. PML and SP100 are covalently modified by the SUMO-1 modifier, which is considered crucial to nuclear body interactions. The encoded protein binds heterochromatin proteins and is thought to play a role in tumorigenesis, immunity, and gene regulation. Alternatively spliced variants have been identified for this gene; one of which encodes a high-mobility group protein.
Specifications
| SP100 | |
| Polyclonal | |
| Unconjugated | |
| SP100 | |
| A430075G10Rik; DKFZp686E07254; FLJ00340; FLJ34579; lysp100b; nuclear antigen Sp100; nuclear autoantigen Sp-100; Nuclear dot-associated Sp100 protein; SP 100; SP100; SP100 nuclear antigen; SP100-HMG nuclear autoantigen; speckled 100 kDa | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6672 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P23497 | |
| SP100 | |
| Synthetic peptide directed towards the middle region of human SP100. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction