missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP100 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11377-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a subnuclear organelle and major component of the PML (promyelocytic leukemia)-SP100 nuclear bodies. PML and SP100 are covalently modified by the SUMO-1 modifier, which is considered crucial to nuclear body interactions. The encoded protein binds heterochromatin proteins and is thought to play a role in tumorigenesis, immunity, and gene regulation. Alternatively spliced variants have been identified for this gene; one of which encodes a high-mobility group protein.Specifications
| SP100 | |
| Polyclonal | |
| Unconjugated | |
| SP100 | |
| Lysp100b, Nuclear autoantigen Sp 100, SP100, SP100 nuclear antigen, Speckled 100 kDa | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6672 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P23497 | |
| SP100 | |
| SP100 Fusion Protein Ag1939 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction