missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SP110 Antibody (OTI4C1), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74306F
This item is not returnable.
View return policy
Description
SP110 Monoclonal specifically detects SP110 in Human samples. It is validated for Immunohistochemistry.
Specifications
| SP110 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| SP110 | |
| Recombinant protein fragment corresponding to amino acids 232-521 of human SP110 (NP_004501) produced in E.coli. | |
| 0.1 mL | |
| 3431.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI4C1 | |
| Immunohistochemistry | |
| FLJ22835, IFI41, IFI75, interferon-induced protein 41, 30kD, Interferon-induced protein 41/75, interferon-induced protein 75, 52kD, IPR1, phosphoprotein 41, phosphoprotein 75, SP110 nuclear body protein, Speckled 110 kDa, Transcriptional coactivator Sp110, VODI | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction