missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sp56 Monoclonal Antibody (7H12)

Description
MA1-10869 detects mouse Sperm Protein sp56 from mouse and hamster samples. MA1-10869 has been successfully used in ELISA, Western blot, and immunohistochemistry applications. The MA1-10869 immunogen is a mixture of native and formaldehyde-zinc fixed mouse sperm purified through a Percoll gradient.
Specifications
Specifications
| Antigen | Sp56 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 7H12 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Zp3r |
| Gene Accession No. | Q60736 |
| Gene Alias | AC122773.1; AV264399; SP56; sperm fertilization protein 56; zona pellucida 3 receptor; zona pellucida sperm-binding protein 3 receptor; Zp3r |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?