Learn More
Invitrogen™ SP6 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA543649
Product Code. 15536162
  • € 528.00 / 100µL
Description
Peptide sequence: DHLAKHMKTH EGAKEEAAGA ASGEGKAGGA VEPPGGKGKR EAEGSVAPSN Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 93%.
SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes.SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SP6
Polyclonal
Unconjugated
Sp6
1110025J03Rik; AA591031; AI592962; EPFN; EPIPROFIN; Klf14; krueppel-like factor 14; Kruppel-like factor 14; LOW QUALITY PROTEIN: transcription factor Sp6; MGC119662; MGC119663; MGC119664; MGC119665; SP6; Sp6 transcription factor; specificity protein 6; trans-acting transcription factor 6; transcription factor Sp6
Rabbit
Affinity chromatography
RUO
80320
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q3SY56
Sp6
Synthetic peptide directed towards the C-terminal of human SP6 (aa 327-376).