Learn More
Invitrogen™ SP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115955
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SP6.
SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes.SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000).
Specifications
| SP6 | |
| Polyclonal | |
| Unconjugated | |
| Sp6 | |
| 1110025J03Rik; AA591031; AI592962; EPFN; EPIPROFIN; Klf14; krueppel-like factor 14; Kruppel-like factor 14; LOW QUALITY PROTEIN: transcription factor Sp6; MGC119662; MGC119663; MGC119664; MGC119665; SP6; Sp6 transcription factor; specificity protein 6; trans-acting transcription factor 6; transcription factor Sp6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80320, 83395 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q3SY56, Q9ESX2 | |
| Sp6 | |
| A synthesized peptide derived from human SP6(Accession Q3SY56), corresponding to amino acid residues P91-S141. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey | |
| Antibody | |
| IgG |