missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPA17 Monoclonal Antibody (OTI1G1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804224S
This item is not returnable.
View return policy
Description
SPA17 Monoclonal antibody specifically detects SPA17 in Human samples. It is validated for Western BlotSpecifications
| SPA17 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15506 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 53340 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI1G1 | |
| Unconjugated | |
| SPA17 | |
| band 34; cancer/testis antigen 22; CT22; SP17; SP17 protein; SP17-1; SPA17; Sperm autoantigenic protein 17; sperm protein 17; Sperm surface protein Sp17; Unknown (protein for MGC:127855); zgc:153721 | |
| Full length human recombinant protein of human SPA17 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction