missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPACA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00 - € 572.00
Specifications
| Antigen | SPACA3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18461111
|
Novus Biologicals
NBP1-89136-25ul |
25 μL |
€ 280.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18250757
|
Novus Biologicals
NBP1-89136 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SPACA3 Polyclonal specifically detects SPACA3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SPACA3 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CT54ALLP17Cancer/testis antigen 54, LYC3Sperm protein reactive with ASA, Lysozyme-like acrosomal sperm-specific secretory protein ALLP-17, Lysozyme-like protein 3, lysozyme-like sperm-specific secretory protein ALLP17, LYZL31700025M08Rik, SLLP1MGC119058, sperm acrosome associated 3, sperm acrosome membrane-associated protein 3, sperm lysozyme like protein 1, Sperm lysozyme-like protein 1, Sperm protein reactive with antisperm antibodies, SPRASA | |
| SPACA3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 124912 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TNNGIFQINSRRWCSNLTPNVPNVCRMYCSDLLNPNLKDTVICAMKITQEPQGLGYWEAWRHHCQGKDLTEWVDGCDF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title