missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPACA9 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82011JF646
This item is not returnable.
View return policy
Description
SPACA9 Polyclonal antibody specifically detects SPACA9 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| SPACA9 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Janelia Fluor 646 | |
| C20orf117, chromosome 20 open reading frame 117, dJ132F21.1, FLJ44670, hypothetical protein LOC140710, KIAA0889, SOGA, suppressor of glucose, autophagy associated 1 | |
| SPACA9 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human SPACA9. The immunogen is located within the last 50 amino acids of SPACA9 . Amino Acid Squence: RLHPSESSWGGEERA | |
| 0.1 mL | |
| Cell Biology | |
| 140710 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction