Learn More
B Braun™ Space PCA Infusion Pump System
Product Code. 10329893
Change view
Click to view available options
Model:
Space PCA Infusion Pump System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
10329893 Space PCA Infusion Pump System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10329893 Supplier B Braun™ Supplier No. 8713030
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Small syringe pump that is big on safety and accuracy in any care setting. B Braun™ Space PCA Infusion Pump System is ideal for operating rooms, adult care, pediatric and emergency settings where ease of use and precise medication administration are critical.
  • Independent Modularity: A microprocessor in each pump helps prevent channel confusion and catastrophic pump failures.
  • Easily Stackable: Vertical stacking takes less space for increased versatility.
    KeyGuard™: Non-numeric keypad with simple arrows to help prevent manual programming errors related to zero vs. decimal entry.
  • Intuitive Universal User Interface on all Space Pumps reduces training required by staff.
  • Extensive Drug Library with customizable hard and soft limits and clinical advisories
    Integrated piston brake helps prevent inadvertent bolus during syringe change.
  • Post-occlusion bolus reduction software.
  • Automatic drive technology ensures accuracy and safety.
  • Horizontal and vertical syringe pump mounting options.
  • Easy-to-learn, intuitive and user friendly 1,500-entry drug library available.
  • Alarms are audible, clearly visible and differentiated using different tones.
  • Strong lightweight pole clamp holds up to three pumps mounted horizontally or vertically.
Specifications
Model Space PCA Infusion Pump System
Applications Operating rooms, adult care, pediatric and emergency settings where ease of use and precise medication administration are critical