missing translation for 'onlineSavingsMsg'
Learn More

Hach™ SPADNS Fluoride Reagent Solution

Product Code. 10149904
Change view
Click to view available options
Range:
0.02 - 2.00mg/L F-
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10149904 0.02 - 2.00mg/L F- Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10149904 Supplier Hach™ Supplier No. 44449

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

USEPA accepted for reporting of drinking and wastewater analysis using Fluoride, SPADNS Method (distillation required).

TRUSTED_SUSTAINABILITY

Specifications

Analytes Fluoride (F-)
Certifications/Compliance EPA compliant
Color Dark red
Composition Hydrochloric acid, Sodium arsenite, Zirconium oxychloride
pH Range <0.5
Volume (Metric) 500 mL
Format Solution
For Use With (Application) Drinking water/wastewater analysis
For Use With (Equipment) All photometers except DR/820
No. of Tests 125
Quantity 500 mL
Range 0.02 - 2.00mg/L F-
Sample Size 10 mL
Storage Requirements Keep containers tightly closed in a dry, cool and well-ventilated place.
Test Method SPADNS
Test Time 1 min
Type Reagent solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • SPADNS REAGENT SOLN
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric acid, Sodium arsenite, 4,5-dihydroxy-3-[(4-sulfophenyl)azo]-2,7-Naphthalenedisulfonic a
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.