missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG16 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97844-100ul
This item is not returnable.
View return policy
Description
SPAG16 Polyclonal antibody specifically detects SPAG16 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SPAG16 | |
| Polyclonal | |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:500-1:2500 | |
| DKFZp666P1710, FLJ22724, FLJ37717, Pf20 protein homolog, PF20MGC87036, sperm associated antigen 16, sperm-associated antigen 16 protein, sperm-associated WD repeat protein, WD repeat domain 29, WDR29 | |
| Produced in rabbits immunized with E. coli-derived Human SPAG16 fragment. | |
| 100 μg | |
| Neuroscience | |
| 79582 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction