missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPAG16 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142540
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Additional Information: This antibody is expected to recognize isoform 1 (NP_078808.3) only. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively.
Spezifikation
| SPAG16 | |
| Polyclonal | |
| Unconjugated | |
| SPAG16 | |
| 4921511D23Rik; 4930524F24Rik; 4930585K05Rik; AV261009; PF20; Pf20 protein homolog; RGD1565062; SPAG16; sperm associated antigen 16; sperm-associated antigen 16 protein; sperm-associated WD repeat protein; spg16; WD repeat domain 29; WDR29 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 79582 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8N0X2 | |
| SPAG16 | |
| Peptide with sequence C-KLHYASYEPTIR. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur